Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Natriumchlorid -Lösung 2 M, in single-use containers, HyClone™ für Bioverfahrenstechnik

Lieferant: Thermo Scientific

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


SH31141.01 SH31141.02 SH31141.03
SH31141.01EA 650 EUR InStock
SH31141.01 SH31141.02 SH31141.03
Natriumchlorid -Lösung 2 M, in single-use containers, HyClone™ für Bioverfahrenstechnik
Natriumchlorid
HyClone™ NaCl solutions in single-use bags offer a convenient format for many upstream and downstream bioprocessing applications.

  • Manufactured in ISO 9001 and ISO 13485 certified facility.
  • Lot-specific certificate of analysis (CoA) for each lot produced.
  • Intended for further manufacturing or research use. Not for diagnostic or therapeutic use.
  • 0.2 µm sterile filtered.
  • Provided in single-use bag containers.
  • Raw material is EP, USP, and JP grade.

These products are manufactured upon order (100 L minimum order quantity). HyClone™ NaCl in single-use bags is well-suited for use with: Cytiva's chromatography resins.
Available in 1 M and 2 M sodium chloride concentrations up to 200 L. Contact us for information around bag configurations or custom bulk formulations.


Formel: NaCl
Storage Temperature: Raumtemperatur
MDL Number: MFCD00003477
CAS-Nummer: 7647-14-5
Order Now
Spezifikation Testergebnisse
Storage Room Temperature as defined by regional Pharmacopeia (15-30°C in US; 15-25°C in EU)
Endotoxin Concentration ≤ 1 EU/mL
Certifications/Compliance Manufactured in a ISO9001 and ISO13485 certified facility
Shelf life 12 Months
For use with Chromatography resins, IC systems, ÄKTA hardware systems
Appearance Clear, colorless solution
Sterility No Growth

 

PRODUKT

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge