100-111-100UG
100-111-10UG
100-111-1MG
100-111-500UG
100-111-50UG
SHBT100-111-100UGEA
327
EUR
InStock
SHBT100-111-100UG
SHBT100-111-10UG
SHBT100-111-1MG
SHBT100-111-500UG
SHBT100-111-50UG
Human Recombinant TNF-A (from E. coli)
Proteine und Peptide
Tumor necrosis factor alpha (TNF-α) is an inflammatory cytokine secreted by macrophages, monocytes, neutrophils, T cells, and NK-cells following stimulation by bacterial lipopolysaccharide (LPS).
- High quality
- Low endotoxin
- Affordable
TNF-α signal activation occurs through two receptors, TNFR1 and TNFR2. TNFR1 is expressed on most cell types, unlike TNFR2, which is expressed mainly on immune cells. TNF-α functions to stimulate phagocytosis in macrophages, chemoattract neutrophils, increase insulin resistance, and induce fever.
Lieferumfang: A certificate of analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
Achtung! Bitte beachten: For research use only.