Human Recombinant TNF-A (from E. coli)
Lieferant: SHENANDOAH BIOTECHNOLOGY
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
100-111-100UG 100-111-10UG 100-111-1MG 100-111-500UG 100-111-50UG
SHBT100-111-100UGEA 327 EUR InStock
SHBT100-111-100UG SHBT100-111-10UG SHBT100-111-1MG SHBT100-111-500UG SHBT100-111-50UG
Human Recombinant TNF-A (from E. coli)
Proteine und Peptide
Tumor necrosis factor alpha (TNF-α) is an inflammatory cytokine secreted by macrophages, monocytes, neutrophils, T cells, and NK-cells following stimulation by bacterial lipopolysaccharide (LPS).
  • High quality
  • Low endotoxin
  • Affordable
TNF-α signal activation occurs through two receptors, TNFR1 and TNFR2. TNFR1 is expressed on most cell types, unlike TNFR2, which is expressed mainly on immune cells. TNF-α functions to stimulate phagocytosis in macrophages, chemoattract neutrophils, increase insulin resistance, and induce fever.
Lieferumfang: A certificate of analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
Achtung! Bitte beachten: For research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge