Human Recombinant HB-EGF (Animal Free) (from E. coli)
Lieferant: SHENANDOAH BIOTECHNOLOGY
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
100-180AF-100U 100-180AF-10UG 100-180AF-1MG 100-180AF-500U 100-180AF-50UG
SHBT100-180AF-100UEA 356 EUR InStock
SHBT100-180AF-100U SHBT100-180AF-10UG SHBT100-180AF-1MG SHBT100-180AF-500U SHBT100-180AF-50UG
Human Recombinant HB-EGF (Animal Free) (from E. coli)
Proteine und Peptide
Heparin-binding EGF-like growth factor (HB-EGF) is a member of the epidermal growth factor (EGF) family and is expressed by monocytes and macrophages. HB-EGF is the predominant growth factor involved in epithelialization during wound healing. HB-EGF signals through the receptor tyrosine kinase ErbB2 to maintain adult heart homeostasis, and promotes cardiac valve development through binding in high affinity to the epidermal growth factor receptor (EGFR). HB-EGF binds the the ErbB4 receptor tyrosine kinase to mediate implantation of the human blastocyst. HB-EGF also functions as a potent mitogen for fibroblasts and smooth muscle cells.
  • High Quality
  • Low Endotoxin
  • Affordable
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.  This product is for research use only and is not for use in humans.  
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
Achtung! Bitte beachten: For research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge