Human Recombinant CNTF (from E. coli)
Lieferant: SHENANDOAH BIOTECHNOLOGY
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
100-59-500UG 100-59-100UG 100-59-1MG 100-59-20UG 100-59-5UG
SHBT100-59-500UGEA 1960 EUR InStock
SHBT100-59-500UG SHBT100-59-100UG SHBT100-59-1MG SHBT100-59-20UG SHBT100-59-5UG
Human Recombinant CNTF (from E. coli)
Proteine und Peptide
Ciliary neurotrophic factor (CNTF) is a neurotrophic factor that promotes the survival of neuronal cell populations, neurite outgrowth, and neurotransmitter synthesis.CNTF also plays an important protective role during nervous system injury.
  • High quality
  • Low endotoxin
  • Affordable
Achtung! Bitte beachten: For research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge