Pig Recombinant IL2 (from E. coli)
Lieferant: SHENANDOAH BIOTECHNOLOGY
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
400-15-5UG 400-15-20UG 400-15-100UG 400-15-500UG 400-15-1MG
SHBT400-15-5UGEA 72.4 EUR InStock
SHBT400-15-5UG SHBT400-15-20UG SHBT400-15-100UG SHBT400-15-500UG SHBT400-15-1MG
Pig Recombinant IL2 (from E. coli)
Proteine und Peptide
Interleukin 2 (IL-2) is an immunomodulatory cytokine that is produced by lymphocytes. IL-2 signals through the IL-2R receptor to induce activated T cell proliferation and promote T cell differentiation. IL-2 also stimulates the proliferation and differentiation of B cells, natural killer cells, monocytes, and macrophages.
  • High Quality
  • Low Endotoxin
  • Affordable
Achtung! Bitte beachten: For research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge