Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

(+)-Limonene 1,2-epoxide cis- und trans-Gemisch ≥97,0% (Summe der Isomere), Sigma-Aldrich®

Lieferant: SIGMAALDRICH

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


17763-100ML 17763-500ML
SIAL17763-100MLEA 160 EUR InStock
SIAL17763-100ML SIAL17763-500ML
(+)-Limonene 1,2-epoxide cis- und trans-Gemisch ≥97,0% (Summe der Isomere), Sigma-Aldrich®
(+)-Limonene 1,2-epoxide
(+)-Limonene 1,2-epoxide is a monoterpene that can be prepared from (+)-limonene has mixture of cis/trans-isomers. It shows potent antitumoral and antinociceptive activity. The main application is used as building block in synthesis of pharmaceutical products, perfumes, cosmetics and food additives.

  • Density: 0,930 g/ml at 20°C (lit.)
  • Refractive index: n20/D 1,467
  • Structure is confirmed by proton NMR spectrum
  • colorless to very faint yellow and colorless to very faint green-yellow, liquid


MW: 152.23 g/mol
CAS-Nummer: 203719-54-4
Order Now

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge