Sample vials, with solid top phenolic screw caps
Lieferant: Sigma-Aldrich
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
27001-U 27002-U 27003 27161 27181 27000 27004 27125-U 27134 27137 27138 27149 27160-U 27171 27172-U 27182 27470-U 27472
SIAL27001-UEA 131 EUR InStock
SIAL27001-U SIAL27002-U SIAL27003 SIAL27161 SIAL27181 SIAL27000 SIAL27004 SIAL27125-U SIAL27134 SIAL27137 SIAL27138 SIAL27149 SIAL27160-U SIAL27171 SIAL27172-U SIAL27182 SIAL27470-U SIAL27472
Sample vials, with solid top phenolic screw caps
Vials
Pre-assembled sample vials with solid-top phenolic screw caps with PTFE or aluminium liner.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge