Non-ionic detergent, Kolliphor® HS 15
Lieferant: Sigma-Aldrich
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
42966-1KG
SIAL42966-1KGEA 246 EUR InStock
SIAL42966-1KG
Non-ionic detergent, Kolliphor® HS 15
Reinigungsmittel Waschmittel und Oberflächenreiniger
Kolliphor HS 15 is a non-ionic detergent obtained by reacting 15 moles of ethylene oxide with 1 mole of 12-hydroxy stearic acid. It's a non-ionic solubiliser and emulsifier.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge