Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Hydroxylamin -Lösung 50% (w/w) in Wasser 99.999%, Sigma-Aldrich®

Lieferant: SIGMAALDRICH

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Gefahr

M-Clarity™ Qualitätsstufe = MQ100

467804-10ML 467804-50ML
SIAL467804-10MLEA 52.6 EUR InStock
SIAL467804-10ML SIAL467804-50ML
Hydroxylamin -Lösung 50% (w/w) in Wasser 99.999%, Sigma-Aldrich®
Hydroxylamin
Hydroxylamine is a weak base that is mainly used as a reducing agent in organic reactions.

Reactant for preparation of:
Prodrug for cardiovascular agent Nω-hydroxy-L-arginine (NOHA, nitric oxide precursor)
Hydroxyaminoguanidines as anti-cancer agents
Nonsteroidal 2,3-dihydroquinoline glucocorticoid receptor agonists with reduced phosphoenolpyruvate caboxykinase (PEPCK) transactivation
Carboxamide derivatives of ofloxacin with improved antimicrobial properties
Analogues of coumarin based TNF-α converting enzyme (TACE) inhibitors
HIV integrase inhibitors


Formel: H₃NO
MW: 33.03 g/mol
Storage Temperature: Raumtemperatur
MDL Number: MFCD00044522
CAS-Nummer: 7803-49-8
EINECS: 232-259-2
UN: 3266
ADR: 8 III
Order Now

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge