Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Hyaluronic acid methacrylate, M.W. 170,000 - 250,000

Lieferant: SIGMAALDRICH

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


M-Clarity™ Qualitätsstufe = MQ100

914800-500MG
SIAL914800-500MGEA 0 EUR InStock
SIAL914800-500MG
Hyaluronic acid methacrylate, M.W. 170,000 - 250,000
Hyaluronic acid methacrylate
Hyaluronic acid (HA) is a linear polysaccharide of alternating D-glucuronic acid and N-acetyl-D-glucosamine found primarily in connective tissues. HA based hydrogels are widely used in tissue engineering, 3D bioprinting, and drug deliery applications. The methacrylate functionalized hyaluronic acid is photo-crosslinkable, and can be used to generate crosslinked hydrogels.


Order Now
Spezifikation Testergebnisse
Appearance (Colour) White to Off White
Appearance (Form) Powder or Chunk(s) or Fibers
1H NMR Spectrum Conforms to Structure
Degree of Functionalisation by HNMR 10 - 30 %
Viscosity 1% in H2O at 25 °C 10 - 20 cps
Molecular Weight (MALLS) 170000 - 250000
ICP Major Analysis, Confirms Sodium Component Confirmed

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge