Tris-EDTA buffer solution, pH 8,0, for molecular biology, BioUltra, Sigma-Aldrich®
Lieferant: SIGMAALDRICH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
93283-100ML 93283-500ML
SIAL93283-100MLEA 51.9 EUR InStock
SIAL93283-100ML SIAL93283-500ML
Tris-EDTA buffer solution, pH 8,0, for molecular biology, BioUltra, Sigma-Aldrich®
Puffer Biologische Puffer
This buffer is 10 mM Tris-HCl and 1 mM disodium EDTA.
  • UV absorption - λ: 260 nm; Amax: 0,04 and λ: 280 nm Amax: 0,01
  • Contains traces of metal cations
Trizma is used in the formulation of buffer solutions in the pH range between 7,5 and 8,5. Tris buffer solutions are widely used in cell and molecular biology for processes such as protein and nucleic acid extraction and purification. Tris-EDTA (TE) buffer solution, pH 8,0 may also be used as a washing buffer.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge