Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Clindamycin hydrochloride, Sigma-Aldrich®

Lieferant: SIGMAALDRICH

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


M-Clarity™ Qualitätsstufe = MQ200

C5269-50MG C5269-100MG C5269-10MG
SIALC5269-50MGEA 503 EUR InStock
SIALC5269-50MG SIALC5269-100MG SIALC5269-10MG
Clindamycin hydrochloride, Sigma-Aldrich®
Clindamycin hydrochloride
Clindamycin is a semi-synthetic, lincosamide antibiotic that is prepared from lincomycin. It inhibits bacterial protein synthesis by hydrogen bond interactions with the 23S rRNA component of the 50S ribosomal subunit thus inducing dissociation of the peptidyl-t-RNA complex. It has antibacterial activity against Gram-positive cocci and antiprotozoal activity against taxoplasma.

  • Effective against anaerobic species
  • Mode of action: protein synthesis
  • Antibiotic activity spectrum: gram-negative bacteria, gram-positive bacteria

Clindamycin is used to study bacterial infections, such as group B streptococcal disease, bacterial resistance and plasma protein binding.

Crystalline

Soluble in water (50 mg/ml)


Formel: C₁₈H₃₃ClN₂O₅S·HCl
MW: 461.45 g/mol
Storage Temperature: Kühlschrank
MDL Number: MFCD07793327
CAS-Nummer: 21462-39-5
Order Now

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge