Scientific Calculator
Lieferant: Sigma-Aldrich
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
C3923-1EA C8474-1EA
SIALC3923-1EAEA 83.6 EUR InStock
SIALC3923-1EA SIALC8474-1EA
Scientific Calculator
Rechner Taschenrechner
Solar powered calculator with protective sliding case.
Functions: Mathematics - Exponential - Trigonometry - Inverse trigonometry - Statistical functions (mean and standard deviation); Angle/time conversions.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge