PUC18 plasmid DNA
Lieferant: SIGMAALDRICH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
D4154-10UG
SIALD4154-10UGEA 199 EUR InStock
SIALD4154-10UG
PUC18 plasmid DNA
Vektoren, Plasmide und Bibliotheken
Plasmids are isolated from host strain E. coli RRI. These plasmids confer ampicillin resistance and complement defects in β-galactosidase in appropriate host strains.
  • Buffered aqueous solution
  • Store at −20 °C
Lieferumfang: Plasmid DNA is supplied in a solution of 10 mM Tris-HCl, pH 8,0, with 1 mM EDTA.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge