Synthetic [D-Ala²]-leucine enkephalin
Lieferant: SIGMAALDRICH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
E5008-5MG
SIALE5008-5MGEA 173 EUR InStock
SIALE5008-5MG
Synthetic [D-Ala²]-leucine enkephalin
Proteine und Peptide
Potent, long-acting Leu-enkephalin analog that is resistant to enzymatic degradation.
  • Amino acid sequence: Tyr-Ala-Gly-Phe-Leu
The enkephalin amid peptide [D-Ala²]-leucine enkephalin ((DADLE), a delta opioid agonist, is a degradation resistant long-acting Leu-enkephalin analog used to study the signaling pathway of delta opioid receptors.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge