Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Perfluorotriamylamine

Lieferant: SIGMAALDRICH

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


F9880-100ML F9880-25ML F9880-SPEC
SIALF9880-100MLEA 920 EUR InStock
SIALF9880-100ML SIALF9880-25ML SIALF9880-SPEC
Perfluorotriamylamine
Perfluorotriamylamine
Fluorinert™ FC-70 has been used to reduce the adherence of large trypsin:N-amidinopiperidine crystals to the well. It has also been used as a fluorocarbon oil for microinjection of oocytes. For use as a high density chase solution in density gradient studies.


Formel: C₁₅F₃₃N
Dichte: 1,93 g/ml (25 °C) (Lit.)
MDL Number: MFCD00042367
CAS-Nummer: 338-84-1
EINECS: 206-421-8
Order Now
Spezifikation Testergebnisse
Appearance (Turbidity) Clear
Appearance (Colour) Colourless
Appearance (Form) Liquid
Boiling Point 212.0 - 218.0 °C
Density 1.910 - 1.950 g/ml
Dielectric Strength ≥35 kV
Distillation Range ≥95 %
Pour Point ≤−15 °C
Residue (100 ml) ≤10.0 µg/ml
Viscosity at 25.0 +/- 0.1 Degrees Celsius 11.0 - 17.0
Vendor Information Confirmed
Recommended Retest Period 3 Years

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge