Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

IGEPAL® CA-630, zäh fließend, Sigma-Aldrich®

Lieferant: SIGMAALDRICH

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Gefahr

M-Clarity™ Qualitätsstufe = MQ200

I3021-50ML I3021-100ML I3021-500ML
SIALI3021-50MLEA 83.3 EUR InStock
SIALI3021-50ML SIALI3021-100ML SIALI3021-500ML
IGEPAL® CA-630, zäh fließend, Sigma-Aldrich®
IGEPAL® CA-630
IGEPAL® CA-630 is a nonionic, non-denaturing detergent suitable for solubilization, isolation and purification of membrane protein complexes. It is chemically indistinguishable from Nonidet P-40.[

IGEPAL® CA-630 has been used:
• As a component of grinding buffer along with lithium dodecyl sulphate, tris-HCl, EDTA, NaCl, sodium deoxycholate, dithiothreitol and thiourea in RNA extraction process
• Along with Tris- HCl finds its application in incubation of Tachyzoites cultured in vero cells
• As a component of nuclear extract buffer D+ containing HEPES, glycerol, KCl and EDTA for gel shift analysis
• As a component of PI buffer along with NaH2PO4 to wash slides in FISH (fluorescence in situ hybridization) procedure


Formel: (C₂H₄O)nC₁₄H₂₂O
Flash-Pt: 251 °C
MDL Number: MFCD00132505
CAS-Nummer: 9002-93-1
UN: 3082
ADR: 9,III
Order Now

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge