Roughness gauge
Lieferant: TQC
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
DC9000
SIMEDC9000EA 5826.17 EUR InStock
SIMEDC9000
Roughness gauge
Beschichtungsprüfgeräte
The (Hull) roughness gauge is a compact system to measure the average Hull roughness (AHR) of sea going vessels, especially for ship operators, ship builders, ship repairers and coating manufacturers.
  • Graphical representations of data and storage in multiple batches
  • Inspection survery reports in Microsoft Excel® using USB-cable and software installed
  • Statistics, time/date and location of series and average hull roughness are calculated and stored
A four direction button operates an intuitive menu on a large display. The neck strap keeps the users' hands free.
The sensor is equipped with three non-slip wheels and a carbide tipped stylus. It is moved over the ship's hull in a horizontal way collecting measurements.
Lieferumfang: Supplied with a neck strap, roughness sensor, sensor cable, calibration plate, thumb drive with software and connection cable for PC.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge