Pocket torch
Lieferant: TQC
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
DI0035
SIMEDI0035EA 52.49 EUR InStock
SIMEDI0035
Pocket torch
Beleuchtung Taschenlampen
This powerful TQC aluminium LED pocket flashlight produces a very impressive 200 lumens and provides a projection distance of 250 m. It is suitable for applications in protective coatings, corrosion control, coating laboratories, paint production, surface finishing, powder coating, decorative coatings and building maintenance.
  • Dynamic switch
  • Shockproof casing
  • Four different modes: High beam, low beam, strobe mode, SOS signal mode
Weight: 73 g
Lieferumfang: Supplied with a holster, a wriststrap, three AAA alkaline batteries and a gift box.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge