Multi-gas detector, GasAlert MicroClip
Lieferant: TQC
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
DM6000
SIMEDM6000EA 860 EUR InStock
SIMEDM6000
Multi-gas detector, GasAlert MicroClip
Gas-Spürsysteme
The GasAlert MicroClip offers simultaneous monitoring of two gasses (O₂, LEL) in a rugged compact case for confined areas.
  • Hazardous materials (HAZMAT) response and other applications requiring gas monitoring
  • One-button operation with automatic calibration
  • Can record and transmit up to ten gas alarm events
Lieferumfang: Supplied with sensors, lithium battery, stainless steel belt clip, calibration hose/cap and manual.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge