Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Surface profile readers, PosiTector® RTR, DeFelsko®

Lieferant: TQC

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

LD5430 LD5431 LD5432 LD5433 LD5434
SIMELD5430EA 975 EUR InStock
SIMELD5430 SIMELD5431 SIMELD5432 SIMELD5433 SIMELD5434
Surface profile readers, PosiTector® RTR, DeFelsko®
Beschichtungsprüfgeräte
These digital spring micrometer measures and records peak to valley surface profile height using replica tape reader and it provides simple, durable, accurate and versatile reading.

  • Solvent, acid, oil, water and dust resistant – weatherproof
  • Continually displays average, standard deviation, min/max and number of readings
  • Storage of 250 readings (Standard) or 100 000 readings (Advanced)
  • Includes PosiSoft suite of solutions for analyzing and reporting data
  • Transfer data via USB to a PC/Mac and also via Bluetooth wireless technology and WiFi technology in advanced models

RTR-H models can measure peak-to-valley surface profile height (HL).

RTR-P models can also measure peak density (Pd). Advanced RTR-P models generate 2D/3D images and surface data files (SDF).

Zertifizierungen: ASTM D4417, NACE RP0287, SSPC PA 17, SSPC-SP5, SP6, SP10, SP11-87T, AS 3894.5, ISO 8503-5 and others.

Lieferumfang: Supplied with stainless steel burnishing tool, five cleaning cards, check shim(s), surface cleaning putty, protective rubber holster with belt clip, wrist strap, nylon carrying case with shoulder strap, protective lens shield, long form certificate of calibration indicating traceability to PTB, USB cable, three alkaline batteries and instructions.

Order Now
 

PRODUKT

TECHNISCHE DATEN

ZUBEHÖR

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge