Adhesion tester, automatic, PosiTest At-A
Lieferant: TQC
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
LD9330
SIMELD9330EA 4060 EUR InStock
SIMELD9330
Adhesion tester, automatic, PosiTest At-A
Adhäsionstester
This automatic adhesion tester measures the adhesion of coatings applied on metal, wood, concrete and similar substrates.
  • Internal memory stores maximum pull-off pressure, rate of pull, test duration, dolly size, pass/fail, nature of fracture and user notes for 100000 pulls in up to 1000 batches
  • Pull rate, pull limit and hold time are user-adjustable
  • Impact and scratch resistant colour touch screen with keypad for operation with or without gloves
  • Built-in rechargeable NiMH battery
The meter is equipped with an electronically controlled hydraulic pump which applies smooth and continuous pull-off pressure. This greatly reduces user effort and the risk of influencing the pulling process. Self-aligning aluminum dollies enable accurate measurements on smooth or uneven surfaces.
Every PosiTest adhesion tester pressure system is calibrated and certified to ±1% accuracy using a NIST traceable load cel. Evaluating measured values on PC is possible by using an adapter and PC-software.
Adhesion strenght: 1 psi (0,01 MPa); ±1% of F.S.
Max. pull force: 500 - 10000 psi (3,5 - 70 MPa), depending on the dolly size
Zertifizierungen: ASTM D4541/D7234, ISO 4624/16276-1, AS/NZS 1580.408.5
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge