Wood moisture meter, mini LIGNO classic+
Lieferant: TQC
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
LI0007
SIMELI0007EA 217 EUR InStock
SIMELI0007
Wood moisture meter, mini LIGNO classic+
Analysatoren Feuchte-Analysatoren
mini LIGNO classic+ is a wood moisture meter that provide four wood groups and 16 single species of wood.
  • Pocket size format
  • Reference scale
Suitable for wood buyers, wood traders, wood processors, saw companies and dry masters.
Lieferumfang: Supplied in a pouch with 5,5 mm and 9 mm pins.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge