pH indicator papers
Lieferant: TQC
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SP2000
SIMESP2000EA 46.2 EUR InStock
SIMESP2000
pH indicator papers
Testpapiere pH-Indikatorstreifen
Universal pH indicator paper on a roll for acid and base (alkaline) test.
  • Fast response time within a minute
  • Easy to read, accurate colour charts
  • High contrast for easy colour chart comparisons
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge