Wedge for measuring reverse-side tolerance
Lieferant: TQC
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SP6000
SIMESP6000EA 69.98 EUR InStock
SIMESP6000
Wedge for measuring reverse-side tolerance
Mess- und Nivelliergeräte
Anodised aluminum wedge for quick measurement of reverse-side tolerance of windows and doors.
  • Tolerance is given in millimetres
  • Also equipped with a measuring scale for optically determining the thickness or glass
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge