Film applicators, spiral type
Lieferant: TQC
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
VF0114 VF0120 VF0119 VF0118 VF0117 VF0115 VF0113 VF0111 VF0110 VF0109 VF0108 VF0107 VF0106 VF0105 VF0104 VF0103 VF0102 VF0101 VF0116 VF0100
SIMEVF0114EA 164.46 EUR InStock
SIMEVF0114 SIMEVF0120 SIMEVF0119 SIMEVF0118 SIMEVF0117 SIMEVF0115 SIMEVF0113 SIMEVF0111 SIMEVF0110 SIMEVF0109 SIMEVF0108 SIMEVF0107 SIMEVF0106 SIMEVF0105 SIMEVF0104 SIMEVF0103 SIMEVF0102 SIMEVF0101 SIMEVF0116 SIMEVF0100
Film applicators, spiral type
Filmaufziehgeräte Filmaufziehgeräte (Farbe und Tinte)
Spiral type applicator with a film width of 250 mm and available in ranges from 4 to 200 μm. Ideal for application of film onto thin materials, such as sheets or plastic. Also works on flexible substrates, and with motorised film applicators. The high grade stainless steel will not be affected by acid and base elements.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge