Prismatic film applicators
Lieferant: TQC
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
VF2163 VF2162 VF2161 VF1837 VF1535 VF1530 VF2164 VF1536 VF1580 VF1581
SIMEVF2163EA 674.18 EUR InStock
SIMEVF2163 SIMEVF2162 SIMEVF2161 SIMEVF1837 SIMEVF1535 SIMEVF1530 SIMEVF2164 SIMEVF1536 SIMEVF1580 SIMEVF1581
Prismatic film applicators
Filmaufziehgeräte Filmaufziehgeräte (Farbe und Tinte)
Prismatic film applicator with four application sides for applying paint films of four different predefined thicknesses. The prism applicators are available in film widths of 50, 75, 80 and 100 mm and are suitable for applying a host of different products onto flat and relatively solid substrates. Made from high grade stainless steel, the prismatic film applicator will not be affected by acid or base elements.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge