Film applicators, Bird-type
Lieferant: TQC
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
VF1541 VF1542 VF1543 VF1544 VF1546 VF1571 VF1570 VF1572 VF1573 VF1574 VF1575 VF1576 VF1545 VF1540
SIMEVF1541EA 355.75 EUR InStock
SIMEVF1541 SIMEVF1542 SIMEVF1543 SIMEVF1544 SIMEVF1546 SIMEVF1571 SIMEVF1570 SIMEVF1572 SIMEVF1573 SIMEVF1574 SIMEVF1575 SIMEVF1576 SIMEVF1545 SIMEVF1540
Film applicators, Bird-type
Filmaufziehgeräte Filmaufziehgeräte (Farbe und Tinte)
Bird-type film applicators feature one flat edge and one application side for applying a pre-defined coating thickness. Bird-type applicator single gap is available in film width 50 and 75 mm and can be customised for applying a host of different products onto flat and relatively solid substrates. All Bird-type film applicators are made from high grade stainless steel and are not affected by acid or base elements.
NOTE: The wet thickness that is obtained cannot be guaranteed. Wet thickness is dependent upon the solids and vehicle content of the wet material, as well as other factors. Film thickness deposited may vary between 40 to 80% of the actual gate clearance of the Bird-type film applicator.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge