Film applicators, multi-functional
Lieferant: TQC
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
VF2174 VF2175 VF2173 VF2172 VF2170 VF2169 VF2168 VF2167
SIMEVF2174EA 556.37 EUR InStock
SIMEVF2174 SIMEVF2175 SIMEVF2173 SIMEVF2172 SIMEVF2170 SIMEVF2169 SIMEVF2168 SIMEVF2167
Film applicators, multi-functional
Filmaufziehgeräte Filmaufziehgeräte (Farbe und Tinte)
Multi-functional film applicator with 4 application sides for applying paint films of 4 different predefined thicknesses. The applicators are available in film widths of 60, 80 mm and one side of the applicator is supplied with a guidance support for straight application. Made from high grade stainless steel, the multi-functional film applicator will not be affected by acid or base elements.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge