Urine containers, SpecTainer™
Lieferant: Simport Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
C566-120CYS
SIMPC566-120CYSEA 226 EUR InStock
SIMPC566-120CYS
Urine containers, SpecTainer™
Klinisches Diagnosezubehör Klinische Probenentnahme und Probenvorbereitung Sammlung und Vorbereitung von Urinproben
PP urine container with tamper evident PE screw cap, ideal for collecting, storing and transporting urine samples.
  • Tamper evident sealing ring for better sample protection
  • Ridges around base offer a better grip during opening and closing
For applications needing the utmost security where sample integrity is of high importance. A great feature of the Spectainer™ I is that it incorporates a unique tamper evident screw cap ensuring peace of mind during transport or storage situations where someone might have tampered with the specimen. For colour coding purposes, place a Capinsert™ on top of closure (see Series T345). Ten different colours are available.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge