This model is a one-piece moulded cassette so no separate lids to manage. It can be opened and closed up to five times guaranteed, securely relocking to avoid specimen loss.
- Unique lateral vents, fluids can access tissue from all 4 sides
- Basket style for small tissue management
- One-piece moulded with front opening
- Smooth 45° angle writing surface
- Available in 11 colours
Moulded from high-density polymer, it keeps specimens safely submerged in liquid.
Available for tissue, biopsy and micro-biopsy. This cassette model is supplied in bulk and also in taped stacks for use with Leica and Sakura Ink Jet printers.
Zertifizierungen: IVDR, CE.