Microarray molds kits, T-Sue™
Lieferant: Simport Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
M473-15 M473-P30P M473-150 M473-36 M473-60 M473-72 M473-P20R M473-P10MA M473-150PC M473-15PC M473-170 M473-170PC M473-24 M473-24PC M473-36PC M473-60PC M473-72PC M473-P15Y M473-P40B
SIMPM473-15EA 1520 EUR InStock
SIMPM473-15 SIMPM473-P30P SIMPM473-150 SIMPM473-36 SIMPM473-60 SIMPM473-72 SIMPM473-P20R SIMPM473-P10MA SIMPM473-150PC SIMPM473-15PC SIMPM473-170 SIMPM473-170PC SIMPM473-24 SIMPM473-24PC SIMPM473-36PC SIMPM473-60PC SIMPM473-72PC SIMPM473-P15Y SIMPM473-P40B
Microarray molds kits, T-Sue™
Mikroarray-Instrumente
T-Sue™ Microarray mold, enabling tissues from many donor blocks to be arrayed on a single slide.
  • Can be reused hundreds of times without losing its flexibility
  • Withstands temperatures between –100 to +250 °C
  • View many different samples on the same slide
  • Array mold is designed to be simple and easy to use
The precast T-Sue™ Microarray paraffin blocks allow users to prepare tissue arrays without needing any specialized equipment. Construct a tissue array block in minutes, simply by punching the donor tissue cores and inserting them into the pre-made paraffin recipient block. No specific technical training or experience is needed.
They are ready to accept cores from a donor block, and offer a consistent core diameter. The cassette is permanently bonded to the block.T-Sue™ punch needles are used for filling recipient tissue microarray paraffin blocks. Internal stylet operates like a retractable pen. Effortless ejection of core specimens.
Tissues can be analyzed in the same conditions enhancing the efficiency of the research. The T-Sue™ Microarray molds allow users to perform TMAs faster while giving excellent results.
Lieferumfang: Each kit includes ONE T-Sue™ array mold and FOUR punch needles with stylet allowing easy removal of tissue cores for insertion.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge