Block mailer, paraffin
Lieferant: Simport Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
M477-6
SIMPM477-6EA 194 EUR InStock
SIMPM477-6
Block mailer, paraffin
Versandtaschen und Transportbehälter
PVC, translucent.
  • Sturdy, secure and easy to use
  • Easy viewing of contents
This mailer can be used for handling and shipping up to 6 blocks as needed and is suitable for all regular models of tissue and biopsy cassettes.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge