Test tube rack, SecuRack™
Lieferant: Simport Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
S510-50O
SIMPS510-50OEA 0 EUR InStock
SIMPS510-50O
Test tube rack, SecuRack™
Gestelle Reagenzglasgestelle
This special 50-place rack will hold your 12×75 mm and 13×100 mm tubes securely in place thanks to silicone tabs surrounding the base of each tube while in the rack. This makes it very convenient to empty tube content before discarding them.
  • Made of high impact polystyrene
  • Silicone tabs secures tubes
  • Alpha numeric grid for each tube position
Also great for holding tubes securely in rack when in a water bath. Units can be anchored laterally to one another, thanks to two screws supplied with each rack.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge