Storage Box for Sample Tubes
Lieferant: Simport Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
T514-2100B T514-581Y T514-225B T514-225G T514-225P T514-225Y T514-2100G T514-2100P T514-2100Y T514-281B T514-281G T514-281P T514-281Y T514-581B T514-581G T514-581P
SIMPT514-2100BEA 269 EUR InStock
SIMPT514-2100B SIMPT514-581Y SIMPT514-225B SIMPT514-225G SIMPT514-225P SIMPT514-225Y SIMPT514-2100G SIMPT514-2100P SIMPT514-2100Y SIMPT514-281B SIMPT514-281G SIMPT514-281P SIMPT514-281Y SIMPT514-581B SIMPT514-581G SIMPT514-581P
Storage Box for Sample Tubes
Boxen Boxen für Reagenzgläser
Cover and base made of polystyrene.
  • Sturdy
  • Stackable
  • Wide variety of economical storage boxes
Printed with a series of numbered squares and the surface accepts the writing with markers by facilitating inventory control.
These storage boxes are designed with a transparent cover that allows the user to see the contents of the box and is keyed to the base to prevent misalignment.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge