Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Transport tubes, selfstanding, non sterile

Lieferant: Simport Scientific

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

T550-10ATPR T550-10AT T552-10ATTP T552-12ATTP T552-30ATTP T552-5ATTP T552-7ATTP
212-9609EA 293 EUR InStock
212-9609 SIMPT550-10AT SIMPT552-10ATTP SIMPT552-12ATTP SIMPT552-30ATTP SIMPT552-5ATTP SIMPT552-7ATTP
Transport tubes, selfstanding, non sterile
Probenbehälter Universalbehälter
Tubes made of non-toxic polypropylene, caps made of polyethylene. Designed for storage and transportation of biological material.

  • Good chemical resistance
  • Good temperature resistance
  • All graduated tubes are in 0,5 ml increments

Tubes provide strength and clarity and exhibit some unique design features. Five sizes are available from 5 to 30 ml. The 212-9609 tube has a white marking area to make sample identification more convenient. They have external threads to provide a smooth and uniform inner surface. A perfect leakproof seal is obtained by the use of a specially designed flexible sealing lip inside the polyethylene closures. Cap also feature a long skirt and a super fast thread design allowing them to be removed or sealed with a single turn.

Bestellinformation: Screw caps have to be ordered separately.

Achtung! Bitte beachten: *Produkte auf dieser Seite sind außerhalb des Anwendungsbereichs der EU Richtlinie 2019/904

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge