Human IL2Ra / CD25 protein (ECD, Fc tag)
Lieferant: SINO BIOLOGICAL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
10165-H02H-100
SINO10165-H02H-100EA 453 EUR InStock
SINO10165-H02H-100
Human IL2Ra / CD25 protein (ECD, Fc tag)
Proteine und Peptide
A DNA sequence encoding the amino acid residues (Met 1-Cys 213) of the human IL2 receptor α chain (NP_000408.1) precursor was expressed with C-terminal fused human IgG1 Fc region.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge