Human ULBP1 / N2DL1 protein (his tag)
Lieferant: SINO BIOLOGICAL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
10679-H08H-100
SINO10679-H08H-100EA 520 EUR InStock
SINO10679-H08H-100
Human ULBP1 / N2DL1 protein (his tag)
Proteine und Peptide
A DNA sequence encoding the mature form of human ULBP1 (NP_079494.1) (Met 1-Gly 216) was expressed, with a polyhistidine tag at the C-terminus.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge