SARS-CoV-2 (2019-nCoV) Nucleocapsid-His Recombinant Protein
Lieferant: SINO BIOLOGICAL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
40588-V07E-100
SINO40588-V07E-100EA 808 EUR InStock
SINO40588-V07E-100
SARS-CoV-2 (2019-nCoV) Nucleocapsid-His Recombinant Protein
Proteine und Peptide
A DNA sequence encoding the SARS-CoV-2 (2019-nCoV) Nucleocapsid Protein (YP_009724397.2) (Met1-Ala419(335Gly/Ala)) was expressed with a polyhistidine tag at the N-terminus.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge