Mouse TNFR1 / CD120a / TNFRSF1A Protein (His Tag)
Lieferant: SINO BIOLOGICAL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
50496-M08H-100
SINO50496-M08H-100EA 0 EUR InStock
SINO50496-M08H-100
Mouse TNFR1 / CD120a / TNFRSF1A Protein (His Tag)
Proteine und Peptide
A DNA sequence encoding the mouse TNFRSF1A (NP_ 035739.2) extracellular domain (Met 1-Ala 212) was expressed, with a polyhistidine tag at the C-terminus.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge