Tissue float bath, MWB
Lieferant: Slee
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
13000500 13000510 13000520
SLEE13000500EA 2800 EUR InStock
SLEE13000500 SLEE13000510 SLEE13000520
Tissue float bath, MWB
Bäder Wasserbäder für Gewebeschnitte
This tissue float bath is designed for stretching of paraffin sections in histology, research and industry laboratories.
  • Black coated bath with stainless steel lid
  • Integrated overheat security
  • Safety thermostat
  • Digital temperature display
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge