Filter tips, Qualitix®
Lieferant: SOCOREX ISBA
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
308.0300FR 3020.0010FRS 3070.0200FRS 3070.1000FRS 3080.0020FRS 3080.0100FRS 3080.0200LFRS 3090.0010FRS
SOCO308.0300FREA 255.5 EUR InStock
SOCO308.0300FR SOCO3020.0010FRS SOCO3070.0200FRS SOCO3070.1000FRS SOCO3080.0020FRS SOCO3080.0100FRS SOCO3080.0200LFRS SOCO3090.0010FRS
Filter tips, Qualitix®
Pipettenspitzen
These tips are made of 100% pure virgin, medical grade PP, fitting not only Socorex® but all major micropipette brands. They include a hydrophobic HDPE filter with 10 µ pore size to block any contaminant.
  • Superior quality, autoclavable
  • Efficient protection against aerosols
  • Lot by lot consistency
  • Certified free of nuclease, endotoxin and ATP
  • Reduced packaging size, less bench space requirement
  • Broad compatibility with other micropipette brands
Beveled tips: Special shape eliminates liquid drop sticking at tip end.
Fine pointed tips: Have a thin, soft end enabling smooth liquid deposit.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge