Cannula with spray nozzle, Dosys™
Lieferant: SOCOREX ISBA
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
370.30500
SOCO370.30500EA 69.5 EUR InStock
SOCO370.30500
Cannula with spray nozzle, Dosys™
Kanülen
This spray nozzle cannula fits on Luer-Lock self-refilling syringes.
Suitable where spraying and nebulising liquid is required.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge