Drawer cabinets 75/66
Lieferant: TRESTON
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
60949101 60949102 60949103 60949105 60949106 60949107 60949108 60949109 60949201 60949202 60949203 60949204 60949205 60949206 60949207 60949208 60949209 60949302 60949303 60949304 60949305 60949306 60949307 60949308 60949309
SOVE60949101EA 0 EUR InStock
SOVE60949101 SOVE60949102 SOVE60949103 SOVE60949105 SOVE60949106 SOVE60949107 SOVE60949108 SOVE60949109 SOVE60949201 SOVE60949202 SOVE60949203 SOVE60949204 SOVE60949205 SOVE60949206 SOVE60949207 SOVE60949208 SOVE60949209 SOVE60949302 SOVE60949303 SOVE60949304 SOVE60949305 SOVE60949306 SOVE60949307 SOVE60949308 SOVE60949309
Drawer cabinets 75/66
Möbel Lagerschränke
Drawer units 75 series designed for storing lightweight items, in environments such as light assembly, inspection, or testing laboratory. These units equipped with base for stand alone version and castors for mobile version.
  • Castors equipped with brakes
  • Maximum loading capacity: 60 kg per drawer
Colour: Grey
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge