Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

S-(Carboxymethyl)-L-cysteine

Lieferant: SPECTRUM CHEMICAL MFG

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


C3071-25KGBL C3071-5KGBL
SPCMC3071-25KGBLEA 0 EUR InStock
SPCMC3071-25KGBL SPCMC3071-5KGBL
S-(Carboxymethyl)-L-cysteine
S-(Carboxymethyl)-L-cysteine
S-Carboxymethyl-L-cysteine, also known as Rhinathiol or Humex, is a mucolytic that minimizes the viscosity of sputum so it can be used to treat chronic obstructive pulmonary disorder and bronchiectasis.

Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Formel: C₅H₉NO₄S
Storage Temperature: Kühlschrank
MDL Number: MFCD00002614
CAS-Nummer: 638-23-3
Order Now

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge