Myrj® S100
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
M1402-500GM
SPCMM1402-500GMEA 135 EUR InStock
SPCMM1402-500GM
Myrj® S100
Myrj® S100
Myrj(R) S100 is a polyethylene glycol ester of stearic acid. In cosmetic and personal care products it functions as a surfactant - cleansing agent in skin creams, conditioners, shampoos, body cleansers and soapless detergents. It cleans the skin and hair by helping water to mix with oil and dirt so that they can be rinsed away. It appears as a cream-colored, waxy solid that is nearly odourless. It is easily soluble in water and acetone.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge