Potassium chloride 99, granular USP
Lieferant: SPECTRUM CHEMICAL MFG
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
PO170-500GM
SPCMPO170-500GMEA 153 EUR InStock
SPCMPO170-500GM
Potassium chloride 99, granular USP
Potassium chloride
Potassium Chloride, Granular, USP is used to treat hypokalemia. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formel: KCl
MW: 74.55 g/mol
Siedepunkt: 1420 °C (1013 hPa)
Melting Pt: 773 °C
Dichte: 1,98 g/cm³ (20 °C)
Storage Temperature: Raumtemperatur
MDL Number: MFCD00011360
CAS-Nummer: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,07621
Order Now
Spezifikation Testergebnisse
Assay 99% Min.
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge