Sodium hydroxide ≥95,0%, Plätzchen FCC
Lieferant: SPECTRUM CHEMICAL MFG
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
S1303-500GM
SPCMS1303-500GMEA 129 EUR InStock
SPCMS1303-500GM
Sodium hydroxide ≥95,0%, Plätzchen FCC
Sodium hydroxide
Sodium Hydroxide, Pellets, FCC is used in the chemical washing and peeling of fruits and vegetables and as a thickening agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
GHS hazard : Corrosion;Exclamation mark
UN1823
Formel: NaOH
MW: 40 g/mol
Siedepunkt: 1390
Melting Pt: 323
Storage Temperature: Raumtemperatur
MDL Number: MFCD00003548
CAS-Nummer: 1310-73-2
UN: 1823
ADR: 8 II
Merck Index: 14,08627
Order Now
Spezifikation Testergebnisse
Assay (Total Alkali As NaOH) 95.0 - 100.5%
Arsenic (As) max. 3 mg/kg
Carbonate (As Na₂CO₃) max. 3.0 %
Insoluble Substances & Organic Matter To Pass Test
Lead (Pb) max. 2 mg/kg
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge