Peristaltic Pumps, KrosFlo®
Lieferant: Spectrum Laboratories
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
ACR1-U2S-01N
SPECACR1-U2S-01NEA 6240 EUR InStock
SPECACR1-U2S-01N
Peristaltic Pumps, KrosFlo®
Pumpen Peristaltikpumpen
Spectrum's KrosFlo Research I (KRI) system combines a digital peristaltic pump and a multi-purpose lab stand to provide ultimate convenience and efficiency for experiments involving fluid circulation and transfers. The easy-to-operate digital KRI pump drives the fluid stream through flexible tubing (to be order separately) while the incorporated stand and included holders secure the tubing, filters, reservoirs and other components in place.
  • Portable compact set-up saves space
  • Accurate and reliable speed control
  • Locking digital keypad and display
  • Easy-to-use and low maintenance pump
  • Speed resolution: 0,1 min-¹
  • Speed accuracy: ±0,25%
Zertifizierungen: ETL, cETL, CE, RoHS and IP 33 safety ratings.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge