Certified colour reference standards for spectrophotometers
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
9423UV95520E 9423UV95100E 401322989841
SPET9423UV95520EEA 2360 EUR InStock
SPET9423UV95520E SPET9423UV95100E SPET401322989841
Certified colour reference standards for spectrophotometers
Standards Farbreferenzen und Spektroskopie-Standards
These colour reference standards can be used with UV-Vis spectroscopy.
  • Each standard includes a calibration certificate
Zertifizierungen: Traceable to ISO/IEC Standard 17025 accredited calibration process.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge